missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BRCA1-associated Protein

Product Code. p-7155809
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16098791

Brand: enQuireBio™ QP9827ye10ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) BRCA1-associated protein, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_006759.3
Gene ID (Entrez) 8315
Name BRCA1-associated Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol BRAP
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence MSVSLVVIRL ELAEHSPVPA GFGFSAAAGE MSDEEIKKTT LASAVACLEG KSPGEKVAII HQHLGRREMT DVIIETMKSN PDELKTTVEE RKSSEASPTA QRSKDHSKEC INAAPDSPSK QLPDQISFFS GNPSVEIVHG IMHLYKTNKM TSLKEDVRRS AMLCILTVPA AMTSHDLMKF VAPFNEVIEQ MKIIRDSTPN QYMVLIKFRA QADADSFYMT CNGRQFNSIE DDVCQLVYVE RAEVLKSEDG ASLPVMDLTE LPKCTVCLER MDESVNGILT TLCNHSFHSQ CLQRWDDTTC PVCRYCQTPE PVEENKCFEC GVQENLWICL ICGHIGCGRY VSRHAYKHFE ETQHTYAMQL TNHRVWDYAG DNYVHRLVAS KTDGKIVQYE CEGDTCQEEK IDALQLEYSY LLTSQLESQR IYWENKIVRI EKDTAEEINN MKTKFKETIE KCDNLEHKLN DLLKEKQSVE RKCTQLNTKV AKLTNELKEE QEMNKCLRAN QVLLQNKLKE EERVLKETCD QKDLQITEIQ EQLRDVMFYL ETQQKINHLP AETRQEIQEG QINIAMASAS SPASSGGSGK LPSRKGRSKR GK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.