missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BTN3A1 / CD277 Protein

A DNA sequence encoding the Homo sapiens (Human) BTN3A1 / CD277, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5729-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927319

  • 476.00€ / 100µg

Estimated Shipment: 25-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001138480.1
BTN3A1 / CD277 Protein
100 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
11119
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BTN3A1
Recombinant Protein
E. coli
QFSVLGPSGP ILAMVGEDAD LPCHLFPTMS AETMELKWVS SSLRQVVNVY ADGKEVEDRQ SAPYRGRTSI LRDGITAGKA ALRIHNVTAS DSGKYLCYFQ DGDFYEKALV ELKVAALGSD LHVDVKGYKD GGIHLECRST GWYPQPQIQW SNNKGENIPT VEAPVVADGV GLYAVAASVI MRGSSGEGVS CTIRSSLLGL EKTASISIAD PFFRSAQRWI AALAG
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions