missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BTN3A3 Protein

A DNA sequence encoding the Homo sapiens (Human) BTN3A3, was expressed in the hosts and tags indicated.

202.00€ - 2765.00€

Specifications

Accession Number NP_001229732.1
Gene ID (Entrez) 10384
Name BTN3A3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15927349
View Documents Promotion Details
enQuireBio™
QP5731-YE-100UG
100 μg
645.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15947349
View Documents Promotion Details
enQuireBio™
QP5731-YE-1MG
1 mg
2765.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15937349
View Documents Promotion Details
enQuireBio™
QP5731-YE-10UG
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15957349
View Documents Promotion Details
enQuireBio™
QP5731-YE-200UG
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15967349
View Documents Promotion Details
enQuireBio™
QP5731-YE-500UG
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15977349
View Documents Promotion Details
enQuireBio™
QP5731-YE-50UG
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001229732.1
BTN3A3 Protein
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
10384
Research Use Only
BTN3A3
Recombinant Protein
Yeast
QFSVLGPSGP ILAMVGEDAD LPCHLFPTMS AETMELRWVS SSLRQVVNVY ADGKEVEDRQ SAPYRGRTSI LRDGITAGKA ALRIHNVTAS DSGKYLCYFQ DGDFYEKALV ELKVAALGSD LHIEVKGYED GGIHLECRST GWYPQPQIKW SDTKGENIPA VEAPVVADGV GLYAVAASVI MRGSSGGGVS CIIRNSLLGL EKTASISIAD PFFRSAQPW
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications