missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CARD18 Protein

A cDNA sequence encoding the CARD18 was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name CARD18 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15936698
View Documents Promotion Details
enQuireBio™
QP11277-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15916698
View Documents Promotion Details
enQuireBio™
QP11277-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15926698
View Documents Promotion Details
enQuireBio™
QP11277-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CARD18 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The LSM3 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl, 1mM DTT and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MADQLLRKKR RIFIHSVGAG TINALLDCLL EDEVISQEDM NKVRDENDTV MDKARVLIDL VTGKGPKSCC KFIKHLCEED PQLASKMGLH
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications