missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Casein kinase I isoform epsilon Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform epsilon, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_001276841.1 |
---|---|
Gene ID (Entrez) | 102800317 |
Name | Casein kinase I isoform epsilon Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978409
|
enQuireBio™
QP5880-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918419
|
enQuireBio™
QP5880-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988409
|
enQuireBio™
QP5880-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15908419
|
enQuireBio™
QP5880-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15928419
|
enQuireBio™
QP5880-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15998409
|
enQuireBio™
QP5880-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
NP_001276841.1 | |
Casein kinase I isoform epsilon Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
102800317 | |
Research Use Only | |
CSNK1E | |
Recombinant Protein | |
Yeast | |
MELRVGNKYR LGRKIGSGSF GDIYLGANIA SGEEVAIKLE CVKTKHPQLH IESKFYKMMQ GGVGIPSIKW CGAEGDYNVM VMELLGPSLE DLFNFCSRKF SLKTVLLLAD QMISRIEYIH SKNFIHRDVK PDNFLMGLGK KGNLVYIIDF GLAKKYRDAR THQHIPYREN KNLTGTARYA SINTHLGIEQ SRRDDLESLG YVLMYFNLGS LPWQGLKAAT KRQKYERISE KKMSTPIEVL CKGYPSEFST YLNFCRSLRF DDKPDYSYLR QLFRNLFHRQ GFSYDYVFDW NMLKFGAARN PEDVDRERRE HEREERMGQL RGSATRALPP GPPTGATANR LRSAAEPVAS TPASRIQPAG NTSPRAISRV DRERKVSMRL HRGAPANVSS SDLTGRQEVS RIPASQTSVP FDHLGK | |
Greater than 90% as determined by SDS-PAGE. |