missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL18 / PARC / MIP4 Protein

Recombinant Protein

2600.00€ - 4240.00€

Especificaciones

Accession Number NP_002979.1
Gene ID (Entrez) 6362
Name CCL18 / PARC / MIP4 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Ver más especificaciones

Productos 2
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
15925789
missing translation for 'viewDocuments' Detalles de la promoción
enQuireBio™
QP5260-500UG
500 μg
2600.00€
500µg
Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915789
missing translation for 'viewDocuments' Detalles de la promoción
enQuireBio™
QP5260-1MG
1 mg
4240.00€
1mg
Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Especificaciones

Especificaciones

NP_002979.1
CCL18 / PARC / MIP4 Protein
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human CCL18 / PARC / MIP4 induces primary human T cell chemotaxis.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
6362
Research Use Only
CCL18
Recombinant Protein
E. coli
AQVGTNKELC CLVYTSWQIP QKFLVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documentos

Documentos

Certificados