missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD117/c-kit Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human CD117/c-kit Protein is derived from Sf 21 (baculovirus). The Recombinant Human CD117/c-kit Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3815 |
| Molecular Weight (g/mol) | 55.7 kDa |
| Name | CD117/c-kit |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human CD117/c-kit protein Gln26-Thr520 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction