Learn More
enQuireBio™ Recombinant Human CDC34 Protein
A cDNA sequence encoding the CDC34 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11363-20ug
Additional Details : Weight : 0.01000kg
Specifications
CDC34 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MARPLVPSSQ KALLLELKGL QEEPVEGFRV TLVDEGDLYN WEVAIFGPPN TYYEGGYFKA RLKFPIDYPY SPPAFRFLTK MWHPNIYETG DVCISILHPP VDDPQSGELP SERWNPTQNV RTILLSVISL LNEPNTFSPA NVDASVMYRK WKESKGKDRE YTDIIRKQVL GTKVDAERDG VKVPTTLAEY CVKTKAPAPD EGSDLFYDDY YEDGEVEEEA DSCFGDDEDD SGTEES | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CDC34 (1 mg/ml) protein solution contains 20mM Tris-Hcl pH 8.0, 10% glycerol, 50mM NaCl and 1mM DTT. |