missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEBP Alpha Protein
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
Specifications
Specifications
| Name | CEBP Alpha Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMGAG KAKKSVDKNS NEYRVRRERN NIAVRKSRDK AKQRNVETQQ KVLELTSDND RLRKRVEQLS RELDTLRGIF RQLPESSLVKAMGNCA |
| Buffer | The protein contains 20mM Tris-HCl pH 7.5, 0.1M NaCl and 5mM b-Mercaptoethanol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction