missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cellular nucleic acid-binding Protein

A DNA sequence encoding the Homo sapiens (Human) Cellular nucleic acid-binding protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5847-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958199

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Specifications

Specifications

NP_001120664.1
Cellular nucleic acid-binding Protein
Research Use Only
CNBP
Recombinant Protein
E. coli
SSNECFKCGR SGHWARECPT GGGRGRGMRS RGRGFQFVSS SLPDICYRCG ESGHLAKDCD LQEDACYNCG RGGHIAKDCK EPKREREQCC YNCGKPGHLA RDCDHADEQK CYSCGEFGHI QKDCTKVKCY RCGETGHVAI NCSKTSEVNC YRCGESGHLA RECTIEATA
Greater than 90% as determined by SDS-PAGE.
7555
10 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers