missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cellular nucleic acid-binding Protein
A DNA sequence encoding the Homo sapiens (Human) Cellular nucleic acid-binding protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5847-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001120664.1 | |
Cellular nucleic acid-binding Protein | |
Research Use Only | |
CNBP | |
Recombinant Protein | |
E. coli | |
SSNECFKCGR SGHWARECPT GGGRGRGMRS RGRGFQFVSS SLPDICYRCG ESGHLAKDCD LQEDACYNCG RGGHIAKDCK EPKREREQCC YNCGKPGHLA RDCDHADEQK CYSCGEFGHI QKDCTKVKCY RCGETGHVAI NCSKTSEVNC YRCGESGHLA RECTIEATA | |
Greater than 90% as determined by SDS-PAGE. |
7555 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |