missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CHMP6 Protein

A cDNA sequence encoding the CHMP6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11422-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927218

  • 125.00€ / 5µg

Estimated Shipment: 28-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

CHMP6 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CHMP6 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 100mM NaCl.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMGNLFG RKKQSRVTEQ DKAILQLKQQ RDKLRQYQKR IAQQLERERA LARQLLRDGR KERAKLLLKK KRYQEQLLDR TENQISSLEA MVQSIEFTQI EMKVMEGLQF GNECLNKMHQ VMSIEEVERI LDETQEAVEY QRQIDELLAG SFTQEDEDAI LEELSAITQE QIELPEVPSE PLPEKIPENV PVKARPRQAE LVAAS
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions