missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CKB Protein

Product Code. p-7148354
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15904228

Brand: enQuireBio™ QP1055210ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CKB was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 1152
Name CKB Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol CKB
Biological Activity The biological activity measured by the enzymatic activity of Creatine phosphokinase procedure No. 45-UV, 1IU-1 micromole creatine phosphate was 854 IU/mg at 37 degrees celsius corresponding to a Specific Activity of 1,171ng/ml.
Product Type Recombinant Protein
Cross Reactivity Human
Species P. pastoris
Protein Tag Untagged
Sequence MPFSNSHNAL KLRFPAEDEF PDLSAHNNHM AKVLTPELYA ELRAKSTPSG FTLDDVIQTG VDNPGHPYIM TVGCVAGDEE SYEVFKDLFD PIIEDRHGGY KPSDEHKTDL NPDNLQGGDD LDPNYVLSSR VRTGRSIRGF CLPPHCSRGE RRAIEKLAVE ALSSLDGDLA GRYYALKSMT EAEQQQLIDD HFLFDKPVSP LLLASGMARD WPDARGIWHN DNKTFLVWVN EEDHLRVISM QKGGNMKEVF TRFCTGLTQI ETLFKSKDYE FMWNPHLGYI LTCPSNLGTG LRAGVHIKLP NLGKHEKFSE VLKRLRLQKR GTGGVDTAAV GGVFDVSNAD RLGFSEVELV QMVVDGVKLL IEMEQRLEQG QAIDDLMPAQ K
Buffer CKBB Human contains 10Mm Bis-Tris-HCl pH 6.0, 50% glycerol, 0.5mM EDTA and 0.5mM DTT.
Purity or Quality Grade Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.