missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKS1B Protein
A cDNA sequence encoding the CKS1B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11438-1mg
Additional Details : Weight : 0.01000kg
Specifications
CKS1B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSHKQIY YSDKYDDEEF EYRHVMLPKD IAKLVPKTHL MSESEWRNLG VQQSQGWVHY MIHEPEPHIL LFRRPLPKKP KK | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CKS1B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 150mM NaCl and 20% glycerol. |