missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CKS1B Protein

A cDNA sequence encoding the CKS1B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11438-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927268

  • 137.00€ / 5µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CKS1B Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSHKQIY YSDKYDDEEF EYRHVMLPKD IAKLVPKTHL MSESEWRNLG VQQSQGWVHY MIHEPEPHIL LFRRPLPKKP KK
Greater than 95% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The CKS1B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 150mM NaCl and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions