missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLIC1 Protein
A cDNA sequence encoding the CLIC1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11447-1mg
Additional Details : Weight : 0.01000kg
Specifications
CLIC1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
GSSHHHHHH SSGLVPRGSH MAEEQPQVEL FVKAGSDGAK IGNCPFSQRL FMVLWLKGVT FNVTTVDTKR RTETVQKLCP GGQLPFLLYG TEVHTDTNKI EEFLEAVLCP PRYPKLAALN PESNTAGLDI FAKFSAYIKN SNPALNDNLE KGLLKALKVL DNYLTSPLPE EVDETSAEDE GVSQRKFLDG NELTLADCNL LPKLHIVQVV CKKYRGFTIP EAFRGVHRYL SNAYAREEFA STCPDDEEIE LAYEQVAKAL K | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CLIC1 Human solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol. |