missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CPSF4 Protein
A DNA sequence encoding the Homo sapiens (Human) CPSF4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5858-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001075028.1 | |
CPSF4 Protein | |
Research Use Only | |
CPSF4 | |
Recombinant Protein | |
E. coli | |
MQEIIASVDH IKFDLEIAVE QQLGAQPLPF PGMDKSGAAV CEFFLKAACG KGGMCPFRHI SGEKTVVCKH WLRGLCKKGD QCEFLHEYDM TKMPECYFYS KFGECSNKEC PFLHIDPESK IKDCPWYDRG FCKHGPLCRH RHTRRVICVN YLVGFCPEGP SCKFMHPRFE LPMGTTEQPP LPQQTQPPAK QRTPQVIGVM QSQNSSAGNR GPRPLEQVTC YKCGEKGHYA NRCTKGHLAF LSGQ | |
Greater than 80% as determined by SDS-PAGE. |
10898 | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |