missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRIPT Protein

A cDNA sequence encoding the CRIPT was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11504-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15907468

  • 212.00€ / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CRIPT Protein
Research Use Only
Recombinant Protein
E. coli
MGSIHHHHHH SSGLVPRGSH MGSMVCEKCE KKLGTVITPD TWKDGARNTT ESGGRKLNEN KALTSKKARF DPYGKNKFST CRICKSSVHQ PGSHYCQGCA YKKGICAMCG KKVLDTKNYK QTSV
Greater than 95% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The CRIPT solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions