missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRYM Protein
A cDNA sequence encoding the CRYM was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | CRYM Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15937528
|
enQuireBio™
QP11524-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15917528
|
enQuireBio™
QP11524-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927528
|
enQuireBio™
QP11524-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CRYM Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CRYM was filtered (0.4microm) and lyophilized from 0.5 mg/ml in 20mM Tris buffer and 50mM NaCl, pH 7.5. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS RVPAFLSAAE VEEHLRSSSL LIPPLETALA NFSSGPEGGV MQPVRTVVPV TKHRGYLGVM PAYSAAEDAL TTKLVTFYED RGITSVVPSH QATVLLFEPS NGTLLAVMDG NVITAKRTAA VSAIATKFLK PPSSEVLCIL GAGVQAYSHY EIFTEQFSFK EVRIWNRTKE NAEKFADTVQ GEVRVCSSVQ EAVAGADVII TVTLATEPIL FGEWVKPGAH INAVGASRPD WRELDDELMK EAVLYVDSQE AALKESGDVL LSGAEIFAEL GEVIKGVKPA HCEKTTVFKS LGMAVEDTVA AKLIYDSWSS GK | |
Greater than 95.0% as determined by SDS-PAGE. |