missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSTA Protein
A cDNA sequence encoding the CSTA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10563-1mg
Additional Details : Weight : 0.01000kg
Specifications
1475 | |
Greater than 95% as determined by reducing SDS-PAGE. | |
Research Use Only | |
CSTA | |
Human | |
His | |
Cystatin-A (1 mg/ml) in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |
CSTA Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MIPGGLSEAK PATPEIQEIV DKVKPQLEEK TNETYGKLEA VQYKTQVVAG TNYYIKVRAG DNKYMHLKVF KSLPGQNEDL VLTGYQVDKN KDDELTGF |