missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CXCL9/MIG Protein

Product Code. 18739376
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18739376

Brand: R&D Systems™ 11106MG050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with mouse CXCR3. The ED50 for this effect is 0.020-0.300 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07325.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 4283
Molecular Weight (g/mol) M.W.-observed: 12-17 kDa, under reducing conditions., M.W.-theroretical: 12 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human CXCL9/MIG protein Thr23-Thr125
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CXCL9/MIG
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.