missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CYP11A1 Protein
A DNA sequence encoding the Homo sapiens (Human) CYP11A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5902-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000772.2 | |
CYP11A1 Protein | |
Research Use Only | |
CYP11A1 | |
Recombinant Protein | |
E. coli | |
ISTRSPRPFN EIPSPGDNGW LNLYHFWRET GTHKVHLHHV QNFQKYGPIY REKLGNVESV YVIDPEDVAL LFKSEGPNPE RFLIPPWVAY HQYYQRPIGV LLKKSAAWKK DRVALNQEVM APEATKNFLP LLDAVSRDFV SVLHRRIKKA GSGNYSGDIS DDLFRFAFES ITNVIFGERQ GMLEEVVNPE AQRFIDAIYQ MFHTSVPMLN LPPDLFRLFR TKTWKDHVAA WDVIFSKADI YTQNFYWELR QKGSVHHDYR GILYRLLGDS KMSFEDIKAN VTEMLAGGVD TTSMTLQWHL YEMARNLKVQ DMLRAEVLAA RHQAQGDMAT MLQLVPLLKA SIKETLRLHP ISVTLQRYLV NDLVLRDYMI PAKTLVQVAI YALGREPTFF FDPENFDPTR WLSKDKNITY FRNLGFGWGV RQCLGRRIAE LEMTIFLINM LENFRVEIQH LSDVGTTFNL ILMPEKPISF TFWPFNQEAT QQ | |
Greater than 90% as determined by SDS-PAGE. |
1583 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |