missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DBI / ACBD1 Protein
A DNA sequence encoding the Homo sapiens (Human) DBI / ACBD1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP1027-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001073331.1 | |
DBI / ACBD1 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1622 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
DBI | |
Recombinant Protein | |
E. coli | |
SQAEFEKAAE EVRHLKTKPS DEEMLFIYGH YKQATVGDIN TERPGMLDFT GKAKWDAWNE LKGTSKEDAM KAYINKVEEL KKKYGI |