missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCX Protein
A DNA sequence encoding the Homo sapiens (Human) DCX, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_000546.2 |
---|---|
Gene ID (Entrez) | 1641 |
Name | DCX Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978679
|
enQuireBio™
QP5920-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15998679
|
enQuireBio™
QP5920-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988679
|
enQuireBio™
QP5920-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15908689
|
enQuireBio™
QP5920-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15918689
|
enQuireBio™
QP5920-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15928689
|
enQuireBio™
QP5920-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
NP_000546.2 | |
DCX Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1641 | |
Research Use Only | |
DCX | |
Recombinant Protein | |
E. coli | |
MKTLPLHSHC TEMQRLLPKL EMLTLGSSFC SLQGEFCQAM DSFTTVSHVG MCEETDASFN VFSPKFQFDR SHCQSLRFHQ NMELDFGHFD ERDKTSRNMR GSRMNGLPSP THSAHCSFYR TRTLQALSNE KKAKKVRFYR NGDRYFKGIV YAVSSDRFRS FDALLADLTR SLSDNINLPQ GVRYIYTIDG SRKIGSMDEL EEGESYVCSS DNFFKKVEYT KNVNPNWSVN VKTSANMKAP QSLASSNSAQ ARENKDFVRP KLVTIIRSGV KPRKAVRVLL NKKTAHSFEQ VLTDITEAIK LETGVVKKLY TLDGKQVTCL HDFFGDDDVF IACGPEKFRY AQDDFSLDEN ECRVMKGNPS ATAGPKASPT PQKTSAKSPG PMRRSKSPAD SANGTSSSQL STPKSKQSPI STPTSPGSLR KHKDLYLPLS LDDSDSLGDS M | |
Greater than 90% as determined by SDS-PAGE. |