missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Desmin Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | Desmin Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MSQAYSSSQR VSSYRRTFGG APGFPLGSPL SSPVFPRAGF GSKGSSSSVT SRVYQVSRTS GGAGGLGSLR ASRLGTTRTP SSYGAGELLD FSLADAVNQE FLTTRTNEKV ELQELNDRFA NYIEKVRFLE QQNAALAAEV NRLKGREPTR VAELYEEELR ELRRQVEVLT NQRARVDVER DNLLDDLQRL KAKLQEEIQL KEEAENNLAA FRADVDAATL ARIDLERRIE SLNEEIAFLK KVHEEEIREL QAQLQEQQVQ VEMDMSKPDL TAALRDIRAQ YETIAAKNIS EAEEWYKSKV SDLTQAANKN NDALRQAKQE MMEYRHQIQS YTCEIDALKG TNDSLMRQMR ELEDRFASEA SGYQDNIARL EEEIRHLKDE MARHLREYQD LLNVKMALDV EIATYRKLLE GEESRINLPI QTYSALNFRE TSPEQRGSEV HTKKTVMIKT IETRDGEVVS EATQQQHEVL |
| Buffer | Desmin was lyophilized from a 1 mg/ml solution containing 30mM Tris-HCl pH 8.0, 9.5M urea, 2mM DTT, 2mM EDTA and 10mM methylammonium chloride. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction