missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Desmoplakin Protein
A DNA sequence encoding the Homo sapiens (Human) Desmoplakin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5952-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001008844.1 | |
Desmoplakin Protein | |
Research Use Only | |
DSP | |
Recombinant Protein | |
E. coli | |
CSDCLMRAEL IVQPELKYGD GIQLTRSREL DECFAQANDQ MEILDSLIRE MRQMGQPCDA YQKRLLQLQE QMRALYKAIS VPRVRRASSK GGGGYTCQSG SGWDEFTKHV TSECLGWMRQ QRAEMDMVAW GVDLASVEQH INSHRGIHNS IGDYRWQLDK IKADLREKSA IYQLEEEYEN LLKASFERMD HLRQLQNIIQ ATSREIMWIN DCEEEELLYD WSD | |
Greater than 80% as determined by SDS-PAGE. |
1832 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |