missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human DLL1 Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
ED50 for this effect is 0.1 to 0.5 μg/mL.
Specifications
Specifications
| Accession Number | O00548 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 28514 |
| Molecular Weight (g/mol) | 88 to 98 kDa, under reducing conditions |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived human DLL1 protein. Human DLL1 (Ser22-Glu537) Accession # O00548IEGRMDPHuman IgG1 (Pro100-Lys330)N-terminus C-terminus |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction