missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA fragmentation factor subunit alpha Protein
A DNA sequence encoding the Homo sapiens (Human) DNA fragmentation factor subunit alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5929-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004392.1 | |
DNA fragmentation factor subunit alpha Protein | |
Research Use Only | |
DFFA | |
Recombinant Protein | |
E. coli | |
MEVTGDAGVP ESGEIRTLKP CLLRRNYSRE QHGVAASCLE DLRSKACDIL AIDKSLTPVT LVLAEDGTIV DDDDYFLCLP SNTKFVALAS NEKWAYNNSD GGTAWISQES FDVDETDSGA GLKWKNVARQ LKEDLSSIIL LSEEDLQMLV DAPCSDLAQE LRQSCATVQR LQHTLQQVLD QREEVRQSKQ LLQLYLQALE KEGSLLSKQE ESKAAFGEEV DAVDTGISRE TSSDVALASH ILTALREKQA PELSLSSQDL ELVTKEDPKA LAVALNWDIK KTETVQEACE WELALRLQQT QSLHSLRSIS ASKASPPGDL QNPKRARQDP T | |
Greater than 80% as determined by SDS-PAGE. |
1676 | |
1 mg | |
Not determined. | |
Full Length of BC007721 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |