missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNAJB4 Protein
A cDNA sequence encoding the DNAJB4 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | DNAJB4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15938008
|
enQuireBio™
QP11676-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918008
|
enQuireBio™
QP11676-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928008
|
enQuireBio™
QP11676-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DNAJB4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DNAJB4 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.2M NaCl, 30% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGKDYYC ILGIEKGASD EDIKKAYRKQ ALKFHPDKNK SPQAEEKFKE VAEAYEVLSD PKKREIYDQF GEEGLKGGAG GTDGQGGTFR YTFHGDPHAT FAAFFGGSNP FEIFFGRRMG GGRDSEEMEI DGDPFSAFGF SMNGYPRDRN SVGPSRLKQD PPVIHELRVS LEEIYSGCTK RMKISRKRLN ADGRSYRSED KILTIEIKKG WKEGTKITFP REGDETPNSI PADIVFIIKD KDHPKFKRDG SNIIYTAKIS LREALCGCSI NVPTLDGRNI PMSVNDIVKP GMRRRIIGYG LPFPKNPDQR GDLLIEFEVS FPDTISSSSK EVLRKHLPAS | |
Greater than 90% as determined by SDS-PAGE. |