missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNAJC15 Protein

A cDNA sequence encoding the DNAJC15 was constructed and used to recombinantly synthesize the protein.

137.00€ - 1815.00€

Specifications

Gene ID (Entrez) 29103
Name DNAJC15 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol DNAJC15
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15928018
View Documents Promotion Details
enQuireBio™
QP11679-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15918018
View Documents Promotion Details
enQuireBio™
QP11679-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15908018
View Documents Promotion Details
enQuireBio™
QP11679-100UG
100 μg
1815.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

29103
Research Use Only
DNAJC15
Human
His
The DNAJC15 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
DNAJC15 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSFRIWKPL EQVITETAKK ISTPSFSSYY KGGFEQKMSR REAGLILGVS PSAGKAKIRT AHRRVMILNH PDKGGSPYVA AKINEAKDLL ETTTKH
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications