missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DYRK2, Isoform 2 Protein
A DNA sequence encoding the Homo sapiens (Human) DYRK2, Isoform 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10058-ba-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003574.1 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
DYRK2 | |
Recombinant Protein | |
Baculovirus | |
MNDHLHVGSH AHGQIQVQQL FEDNSNKRTV LTTQPNGLTT VGKTGLPVVP ERQLDSIHRR QGSSTSLKSM EGMGKVKATP MTPEQAMKQY MQKLTAFEHH EIFSYPEIYF LGLNAKKRQG MTGGPNNGGY DDDQGSYVQV PHDHVAYRYE VLKVIGKGSF GQVVKAYDHK VHQHVALKMV RNEKRFHRQA AEEIRILEHL RKQDKDNTMN VIHMLENFTF RNHICMTFEL LSMNLYELIK KNKFQGFSLP LVRKFAHSIL QCLDALHKNR IIHCDLKPEN ILLKQQGRSG IKVIDFGSSC YEHQRVYTYI QSRFYRAPEV ILGARYGMPI DMWSLGCILA ELLTGYPLLP GEDEGDQLAC MIELLGMPSQ KLLDASKRAK NFVSSKGYPR YCTVTTLSDG SVVLNGGRSR RGKLRGPPES REWGNALKGC DDPLFLDFLK QCLEWDPAVR MTPGQALRHP WLRRRLPKPP TGEKTSVKRI TESTGAITSI SKLPPPSSSA SKLRTNLAQM TDANGNIQQR TVLPKLVS |
DYRK2, Isoform 2 Protein | |
20 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |