missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ECHS1 Protein

A cDNA sequence encoding the ECHS1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10581-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15914318

  • 212.00€ / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1892
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 150 units/mg, and is defined as the amount of enzyme that hydrolyzes 1.0 umole of crotonoyl-CoA to hydroxybutyryl-CoA per minute per minute at pH 7.5 at 25C.
Human
His
ECHS1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 20% Glycerol and 100mM NaCl.
ECHS1 Protein
Research Use Only
ECHS1
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MASGANFEYI IAEKRGKNNT VGLIQLNRPK ALNALCDGLI DELNQALKIF EEDPAVGAIV LTGGDKAFAA GADIKEMQNL SFQDCYSSKF LKHWDHLTQV KKPVIAAVNG YAFGGGCELA MMCDIIYAGE KAQFAQPEIL IGTIPGAGGT QRLTRAVGKS LAMEMVLTGD RISAQDAKQA GLVSKICPVE TLVEEAIQCA EKIASNSKIV VAMAKESVNA AFEMTLTEGS KLEKKLFYST FATDDRKEGM TAFVEKRKAN FKDQ
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions