missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EEF1E1 / AIMP3 / p18 Protein

A DNA sequence encoding the Homo sapiens (Human) EEF1E1 / AIMP3 / p18, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5961-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928959

  • 2385.00€ / 1mg
Estimated Shipment:
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001129122.1
EEF1E1 / AIMP3 / p18 Protein
Research Use Only
EEF1E1
Recombinant Protein
E. coli
AAAAELSLLE KSLGLSKGNK YSAQGERQIP VLQTNNGPSL TGLTTIAAHL VKQANKEYLL GSTAEEKAIV QQWLEYRVTQ VDGHSSKNDI HTLLKDLNSY LEDKVYLTGY NFTLADILLY YGLHRFIVDL TVQEKEKYLN VSRWFCHIQH YPGIRQHLSS VVFIKNRLYT NSH
Greater than 90% as determined by SDS-PAGE.
9521
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions