missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF-5A / EIF5 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF-5A / EIF5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5976-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001960.2 | |
EIF-5A / EIF5 Protein | |
Research Use Only | |
EIF5 | |
Recombinant Protein | |
E. coli | |
MSVNVNRSVS DQFYRYKMPR LIAKVEGKGN GIKTVIVNMV DVAKALNRPP TYPTKYFGCE LGAQTQFDVK NDRYIVNGSH EANKLQDMLD GFIKKFVLCP ECENPETDLH VNPKKQTIGN SCKACGYRGM LDTHHKLCTF ILKNPPENSD SGTGKKEKEK KNRKGKDKEN GSVSSSETPP PPPPPNEINP PPHTMEEEED DDWGEDTTEE AQRRRMDEIS DHAKVLTLSD DLERTIEERV NILFDFVKKK KEEGVIDSSD KEIVAEAERL DVKAMGPLVL TEVLFNEKIR EQIKKYRRHF LRFCHNNKKA QRYLLHGLEC VVAMHQAQLI SKIPHILKEM YDADLLEEEV IISWSEKASK KYVSKELAKE IRVKAEPFIK WLKEAEEESS GGEEEDEDEN IEVVYSKAAS VPKVETVKSD NKDDDIDIDA I | |
Greater than 90% as determined by SDS-PAGE. |
1983 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |