missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF1AY Protein

A cDNA sequence encoding the EIF1AY was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name EIF1AY Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15998288
View Documents Promotion Details
enQuireBio™
QP11764-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15978288
View Documents Promotion Details
enQuireBio™
QP11764-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15988288
View Documents Promotion Details
enQuireBio™
QP11764-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

EIF1AY Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The EIF1AY protein solution (0.5 mg/1ml) is formulated in 20 mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSSMNPEYDY LFKLLLIGDS GVGKSCLLLR FADDTYTESY ISTIGVDFKI RTIELDGKTI KLQIWDTAGQ ERFRTITSSY YRGAHGIIVV YDVTDQESFN NVKQWLQEID RYASENVNKL LVGNKCDLTT KKVVDYTTAK EFADSLGIPF LETSAKNATN VEQSFMTMAA EIKKRMGPGA TAGGAEKSNV KIQSTPVKQS GGGCC
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications