missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF2AK2 Protein

A DNA sequence encoding the Homo sapiens (Human) EIF2AK2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5969-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15919019

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Specifications

Specifications

NP_001129123.1
EIF2AK2 Protein
Research Use Only
EIF2AK2
Recombinant Protein
E. coli
AGDLSAGFFM EELNTYRQKQ GVVLKYQELP NSGPPHDRRF TFQVIIDGRE FPEGEGRSKK EAKNAAAKLA VEILNKEKKA VSPLLLTTTN SSEGLSMGNY IGLINRIAQK KRLTVNYEQC ASGVHGPEGF HYKCKMGQKE YSIGTGSTKQ EAKQLAAKLA YLQILSEETS VKSDYLSSGS FATTCESQSN SLVTSTLASE SSSEGDFSAD TSEINSNSDS LNSSSLLMNG LRNNQRKAKR SLAPRFDLPD MKETKYTVDK RFGMDFKEIE LIGSGGFGQV FKAKHRIDGK TYVIKRVKYN NEKAEREVKA LAKLDHVNIV HYNGCWDGFD YDPETSDDSL ESSDYDPENS KNSSRSKTKC LFIQMEFCDK GTLEQWIEKR RGEKLDKVLA LELFEQITKG VDYIHSKKLI HRDLKPSNIF LVDTKQVKIG DFGLVTSLKN DGKRTRSKGT LRYMSPEQIS SQDYGKEVDL YALGLILAEL LHVCDTAFET SKFFTDLRDG IISDIFDKKE KTLLQKLLSK KPEDRPNTSE ILRTLTVWKK SPEKNERHTC
Greater than 90% as determined by SDS-PAGE.
5610
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions