missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF4EBP1 Protein

A cDNA sequence encoding the EIF4EBP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11773-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968318

  • 137.00€ / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

EIF4EBP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSGGSSCSQT PSRAIPATRR VVLGDGVQLP PGDYSTTPGG TLFSTTPGGT RIIYDRKFLM ECRNSPVTKT PPRDLPTIPG VTSPSSDEPP MEASQSHLRN SPEDKRAGGE ESQFEMDI
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The EIF4EBP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions