missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF4EBP3 Protein

A DNA sequence encoding the Homo sapiens (Human) EIF4EBP3, was expressed in the hosts and tags indicated.

160.00€ - 2385.00€

Specifications

Accession Number NP_003723.1
Gene ID (Entrez) 8637
Name EIF4EBP3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15929059
View Documents Promotion Details
enQuireBio™
QP5974-EC-100UG
100 μg
530.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15949059
View Documents Promotion Details
enQuireBio™
QP5974-EC-1MG
1 mg
2385.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15939059
View Documents Promotion Details
enQuireBio™
QP5974-EC-10UG
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15959059
View Documents Promotion Details
enQuireBio™
QP5974-EC-200UG
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15969059
View Documents Promotion Details
enQuireBio™
QP5974-EC-500UG
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15979059
View Documents Promotion Details
enQuireBio™
QP5974-EC-50UG
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_003723.1
EIF4EBP3 Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
8637
Research Use Only
EIF4EBP3
Recombinant Protein
E. coli
MSTSTSCPIP GGRDQLPDCY STTPGGTLYA TTPGGTRIIY DRKFLLECKN SPIARTPPCC LPQIPGVTTP PTAPLSKLEE LKEQETEEEI PDDAQFEMDI
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications