missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF5A Protein

A cDNA sequence encoding the EIF5A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11775-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938328

  • 212.00€ / 20µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

EIF5A Protein
Research Use Only
Recombinant Protein
E. coli
MADDLDFETG DAGASATFPM QCSALRKNGF VVLKGRPCKI VEMSTSKTGK HGHAKVHLVG IDIFTGKKYE DICPSTHNMD VPNIKRNDFQ LIGIQDGYLS LLQDSGEVRE DLRLPEGDLG KEIEQKYDCG EEILITVLSA MTEEAAVAIK AMAK
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The EIF5A protein solution (1 mg/ml) contains 50mM Tris-HCl, pH 7.5 and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions