missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF5A Protein
A cDNA sequence encoding the EIF5A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11775-1mg
Additional Details : Weight : 0.01000kg
Specifications
EIF5A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MADDLDFETG DAGASATFPM QCSALRKNGF VVLKGRPCKI VEMSTSKTGK HGHAKVHLVG IDIFTGKKYE DICPSTHNMD VPNIKRNDFQ LIGIQDGYLS LLQDSGEVRE DLRLPEGDLG KEIEQKYDCG EEILITVLSA MTEEAAVAIK AMAK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The EIF5A protein solution (1 mg/ml) contains 50mM Tris-HCl, pH 7.5 and 10% Glycerol. |