missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ELP5 Protein
A cDNA sequence encoding the ELP5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11780-10ug
Additional Details : Weight : 0.01000kg
Specifications
ELP5 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ELP5 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
Greater than 80.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTPSEGA RAGTGRELEM LDSLLALGGL VLLRDSVEWE GRSLLKALVK KSALCGEQVH ILGCEVSEEE FREGFDSDIN NRLVYHDFFR DPLNWSKTEE AFPGGPLGAL RAMCKRTDPV PVTIALDSLS WLLLRLPCTT LCQVLHAVSH QDSCPGDSSS VGKVSVLGLL HEELHGPGPV GALSSLAQTE VTLGGTMGQA SAHILCRRPR QRPTDQTQWF SILPDFSLDL QEGPSVESQP YSDPHIPPVD PTTHLTFNLH LSKKEREARD SLILPFQFSS EKQQALLRPR PGQATSHIFY EPDAYDDLDQ EDPDDDLDI |