missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Endostatin Protein

A cDNA sequence encoding the Endostatin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10588-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924338

  • 189.00€ / 100µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Endostatin Protein
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The recombinant Human Endostatin is formulated with 30mM Sodium acetate buffer and Mannitol, pH 5.5.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
The activity was found to be 16,000IU/mg.
Human
Untagged
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions