missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EXOSC10 Protein

A cDNA sequence encoding the EXOSC10 was constructed and used to recombinantly synthesize the protein.

137.00€ - 5300.00€

Specifications

Name EXOSC10 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15938478
View Documents Promotion Details
enQuireBio™
QP11818-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15928478
View Documents Promotion Details
enQuireBio™
QP11818-20UG
20 μg
212.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15918478
View Documents Promotion Details
enQuireBio™
QP11818-1MG
1 mg
5300.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

EXOSC10 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
EXOSC10 is supplied at a concentration of 0.8 mg/ml in 20mM HEPES, pH 7.5, 100mM NaCl, 0.01mM EDTA, 0.02% SDS.
Research Use Only
Recombinant Protein
Baculovirus
MAPPSTREPR VLSATSATKS DGEMVLPGFP DADSFVKFAL GSVVAVTKAS GGLPQFGDEY DFYRSFPGFQ AFCETQGDRL LQCMSRVMQY HGCRSNIKDR SKVTELEDKF DLLVDANDVI LERVGILLDE ASGVNKNQQP VLPAGLQVPK TVVSSWNRKA AEYGKKAKSE TFRLLHAKNI IRPQLKFREK IDNSNTPFLP KIFIKPNAQK PLPQALSKER RERPQDRPED LDVPPALADF IHQQRTQQVE QDMFAHPYQY ELNHFTPADA VLQKPQPQLY RPIEETPCHF ISSLDELVEL NEKLLNCQEF AVDLEHHSYR SFLGLTCLMQ ISTRTEDFII DTLELRSDMY ILNESLTDPA IVKVFHGADS DIEWLQKDFG LYVVNMFDTH QAARLLNLGR HSLDHLLKLY CNVDSNKQYQ LADWRIRPLP EEMLSYARDD THYLLYIYDK MRLEMWERGN GQPVQLQVVW QRSRDICLKK FIKPIFTDES YLELYRKQKK HLNTQQLTAF QLLFAWRDKT ARREDESYGY VLPNHMMLKI AEELPKEPQG IIACCNPVPP LVRQQINEMH LLIQQAREMP LLKSEVAAGV KKSGPLPSAE RLENVLFGPH DCSHAPPDGY PIIPTSGSVP VQKQASLFPD EKEDNLLGTT CLIATAVITL FNEPSAEDSK KGPLTVAQKK AQNIMESFEN PFRMFLPSLG HRAPVSQAAK FDPSTKIYEI SNRWKLAQVQ VQKDSKEAVK KKAAEQTAAR EQAKEACKAA AEQAISVRQQ VVLENAAKKR ERATSDPRTT EQKQEKKRLK ISKKPKDPEP PEKEFTPYDY SQSDFKAFAG NSKSKVSSQF DPNKQTPSGK KCIAAKKIKQ SVGNKSMSFP TGKSDRGFRY NWPQR
Greater than 70% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Certificates