missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC8 Protein
A cDNA sequence encoding the EXOSC8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11823-1mg
Additional Details : Weight : 0.01000kg
Specifications
EXOSC8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAGFKT VEPLEYYRRF LKENCRPDGR ELGEFRTTTV NIGSISTADG SALVKLGNTT VICGVKAEFA APSTDAPDKG YVVPNVDLPP LCSSRFRSGP PGEEAQVASQ FIADVIENSQ IIQKEDLCIS PGKLVWVLYC DLICLDYDGN ILDACTFALL AALKNVQLPE VTINEETALA EVNLKKKSYL NIRTHPVATS FAVFDDTLLI VDPTGEEEHL ATGTLTIVMD EEGKLCCLHK PGGSGLTGAK LQDCMSRAVT RHKEVKKLMD EVIKSMKPK | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EXOSC8 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 50% glycerol, 0.2M NaCl and 1mM DTT. |