missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC8 Protein
A cDNA sequence encoding the EXOSC8 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | EXOSC8 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15988488
|
enQuireBio™
QP11823-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968488
|
enQuireBio™
QP11823-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978488
|
enQuireBio™
QP11823-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
EXOSC8 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EXOSC8 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 50% glycerol, 0.2M NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAGFKT VEPLEYYRRF LKENCRPDGR ELGEFRTTTV NIGSISTADG SALVKLGNTT VICGVKAEFA APSTDAPDKG YVVPNVDLPP LCSSRFRSGP PGEEAQVASQ FIADVIENSQ IIQKEDLCIS PGKLVWVLYC DLICLDYDGN ILDACTFALL AALKNVQLPE VTINEETALA EVNLKKKSYL NIRTHPVATS FAVFDDTLLI VDPTGEEEHL ATGTLTIVMD EEGKLCCLHK PGGSGLTGAK LQDCMSRAVT RHKEVKKLMD EVIKSMKPK | |
Greater than 90% as determined by SDS-PAGE. |