missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human F8 Protein

Product Code. 15924358
Click to view available options
Quantity:
1000 IU
250 IU
500 IU
Unit Size:
1000 units
250 units
500 units
This item is not returnable. View return policy

Product Code. 15924358

Brand: enQuireBio™ QP10594500IU

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the F8 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 2157
Name F8 Protein
Quantity 500 IU
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol F8
Biological Activity The specific activity was found to be 7,058 IU/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Untagged
Sequence MQIELSTCFF LCLLRFCFSA TRRYYLGAVE LSWDYMQSDL GELPVDARFP PRVPKSFPFN TSVVYKKTLF VEFTDHLFNI AKPRPPWMGL LGPTIQAEVY DTVVITLKNM ASHPVSLHAV GVSYWKASEG AEYDDQTSQR EKEDDKVFPG GSHTYVWQVL KENGPMASDP LCLTYSYLSH VDLVKDLNSG LIGALLVCRE GSLAKEKTQT LHKFILLFAV FDEGKSWHSE TKNSLMQDRD AASARAWPKM HTVNGYVNRS LPGLIGCHRK SVYWHVIGMG TTPEVHSIFL EGHTFLVRNH RQASLEISPI TFLTAQTLLM DLGQFLLFCH ISSHQHDGME AYVKVDSCPE EPQLRMKNNE EAEDYDDDLT DSEMDVVRFD DDNSPSFIQI RSVAKKHPKT WVHYIAAEEE DWDYAPLVLA PDDRSYKSQY LNNGPQRIGR KYKKVRFMAY TDETFKTREA IQHESGILGP LLYGEVGDTL LIIFKNQASR PYNIYPHGIT DVRPLYSRRL PKGVKHLKDF PILPGEIFKY KWTVTVEDGP TKSDPRCLTR YYSSFVNMER DLASGLIGPL LICYKESVDQ RGNQIMSDKR NVILFSVFDE NRSWYLTENI QRFLPNPAGV QLEDPEFQAS NIMHSINGYV FDSLQLSVCL HEVAYWYILS IGAQTDFLSV FFSGYTFKHK MVYEDTLTLF PFSGETVFMS MENPGLWILG CHNSDFRNRG MTALLKVSSC DKNTGDYYED SYEDISAYLL SKNNAIEPRS FSQNSRHPST RQKQFNATTI PENDIEKTDP WFAHRTPMPK IQNVSSSDLL MLLRQSPTPH GLSLSDLQEA KYETFSDDPS PGAIDSNNSL SEMTHFRPQL HHSGDMVFTP ESGLQLRLNE KLGTTAATEL KKLDFKVSST SNNLISTIPS DNLAAGTDNT SSLGPPSMPV HYDSQLDTTL FGKKSSPLTE SGGPLSLSEE NNDSKLLESG LMNSQESSWG KNVSSTESGR LFKGKRAHGP ALLTKDNALF KVSISLLKTN KTSNNSATNR KTHIDGPSLL IENSPSVWQN ILESDTEFKK VTPLIHDRML MDKNATALRL NHMSNKTTSS KNMEMVQQKK EGPIPPDAQN PDMSFFKMLF LPESARWIQR THGKNSLNSG QGPSPKQLVS LGPEKSVEGQ NFLSEKNKVV VGKGEFTKDV GLKEMVFPSS RNLFLTNLDN LHENNTHNQE KKIQEEIEKK ETLIQENVVL PQIHTVTGTK NFMKNLFLLS TRQNVEGSYD GAYAPVLQDF RSLNDSTNRT KKHTAHFSKK GEEENLEGLG NQTKQIVEKY ACTTRISPNT SQQNFVTQRS KRALKQFRLP LEETELEKRI IVDDTSTQWS KNMKHLTPST LTQIDYNEKE KGAITQSPLS DCLTRSHSIP QANRSPLPIA KVSSFPSIRP IYLTRVLFQD NSSHLPAASY RKKDSGVQES SHFLQGAKKN NLSLAILTLE MTGDQREVGS LGTSATNSVT YKKVENTVLP KPDLPKTSGK VELLPKVHIY QKDLFPTETS NGSPGHLDLV EGSLLQGTEG AIKWNEANRP GKVPFLRVAT ESSAKTPSKL LDPLAWDNHY GTQIPKEEWK SQEKSPEKTA FKKKDTILSL NACESNHAIA AINEGQNKPE IEVTWAKQGR TERLCSQNPP VLKRHQREIT RTTLQSDQEE IDYDDTISVE MKKEDFDIYD EDENQSPRSF QKKTRHYFIA AVERLWDYGM SSSPHVLRNR AQSGSVPQFK KVVFQEFTDG SFTQPLYRGE LNEHLGLLGP YIRAEVEDNI MVTFRNQASR PYSFYSSLIS YEEDQRQGAE PRKNFVKPNE TKTYFWKVQH HMAPTKDEFD CKAWAYFSDV DLEKDVHSGL IGPLLVCHTN TLNPAHGRQV TVQEFALFFT IFDETKSWYF TENMERNCRA PCNIQMEDPT FKENYRFHAI NGYIMDTLPG LVMAQDQRIR WYLLSMGSNE NIHSIHFSGH VFTVRKKEEY KMALYNLYPG VFETVEMLPS KAGIWRVECL IGEHLHAGMS TLFLVYSNKC QTPLGMASGH IRDFQITASG QYGQWAPKLA RLHYSGSINA WSTKEPFSWI KVDLLAPMII HGIKTQGARQ KFSSLYISQF IIMYSLDGKK WQTYRGNSTG TLMVFFGNVD SSGIKHNIFN PPIIARYIRL HPTHYSIRST LRMELMGCDL NSCSMPLGME SKAISDAQIT ASSYFTNMFA TWSPSKARLH LQGRSNAWRP QVNNPKEWLQ VDFQKTMKVT GVTTQGVKSL LTSMYVKEFL ISSSQDGHQW TLFFQNGKVK VFQGNQDSFT PVVNSLDPPL LTRYLRIHPQ SWVHQIALRM EVLGCEAQDL Y
Buffer Each IU was lyophilized from a solution containing 30ug PEG, 1.6ng VWF, 4ug calcium, and 0.25 mg HSA.
Purity or Quality Grade Greater than 97.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.