missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F8 Protein
Click to view available options
Quantity:
1000 IU
250 IU
500 IU
Unit Size:
1000 units
250 units
500 units
Specifications
Specifications
| Gene ID (Entrez) | 2157 |
| Name | F8 Protein |
| Quantity | 1000 IU |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Gene Symbol | F8 |
| Biological Activity | The specific activity was found to be 7,058 IU/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction