missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F8 Protein
A cDNA sequence encoding the F8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10594-250IU
Additional Details : Weight : 0.01000kg
Specifications
2157 | |
250 IU | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was found to be 7,058 IU/mg. | |
Human | |
Untagged | |
Each IU was lyophilized from a solution containing 30ug PEG, 1.6ng VWF, 4ug calcium, and 0.25 mg HSA. |
F8 Protein | |
Research Use Only | |
F8 | |
Recombinant Protein | |
Human | |
MQIELSTCFF LCLLRFCFSA TRRYYLGAVE LSWDYMQSDL GELPVDARFP PRVPKSFPFN TSVVYKKTLF VEFTDHLFNI AKPRPPWMGL LGPTIQAEVY DTVVITLKNM ASHPVSLHAV GVSYWKASEG AEYDDQTSQR EKEDDKVFPG GSHTYVWQVL KENGPMASDP LCLTYSYLSH VDLVKDLNSG LIGALLVCRE GSLAKEKTQT LHKFILLFAV FDEGKSWHSE TKNSLMQDRD AASARAWPKM HTVNGYVNRS LPGLIGCHRK SVYWHVIGMG TTPEVHSIFL EGHTFLVRNH RQASLEISPI TFLTAQTLLM DLGQFLLFCH ISSHQHDGME AYVKVDSCPE EPQLRMKNNE EAEDYDDDLT DSEMDVVRFD DDNSPSFIQI RSVAKKHPKT WVHYIAAEEE DWDYAPLVLA PDDRSYKSQY LNNGPQRIGR KYKKVRFMAY TDETFKTREA IQHESGILGP LLYGEVGDTL LIIFKNQASR PYNIYPHGIT DVRPLYSRRL PKGVKHLKDF PILPGEIFKY KWTVTVEDGP TKSDPRCLTR YYSSFVNMER DLASGLIGPL LICYKESVDQ RGNQIMSDKR NVILFSVFDE NRSWYLTENI QRFLPNPAGV QLEDPEFQAS NIMHSINGYV FDSLQLSVCL HEVAYWYILS IGAQTDFLSV FFSGYTFKHK MVYEDTLTLF PFSGETVFMS MENPGLWILG CHNSDFRNRG MTALLKVSSC DKNTGDYYED SYEDISAYLL SKNNAIEPRS FSQNSRHPST RQKQFNATTI PENDIEKTDP WFAHRTPMPK IQNVSSSDLL MLLRQSPTPH GLSLSDLQEA KYETFSDDPS PGAIDSNNSL SEMTHFRPQL HHSGDMVFTP ESGLQLRLNE KLGTTAATEL KKLDFKVSST SNNLISTIPS | |
Greater than 97.0% as determined by SDS-PAGE. |