missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FABP1 Protein

A cDNA sequence encoding the FABP1 was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name FABP1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15998508
View Documents Promotion Details
enQuireBio™
QP11830-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15978508
View Documents Promotion Details
enQuireBio™
QP11830-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15988508
View Documents Promotion Details
enQuireBio™
QP11830-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

FABP1 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 7.5.
Research Use Only
Recombinant Protein
E. coli
MSFSGKYQLQ SQENFEAFMK AIGLPEELIQ KGKDIKGVSE IVQNGKHFKF TITAGSKVIQ NEFTVGEECE LETMTGEKVK TVVQLEGDNK LVTTFKNIKS VTELNGDIIT NTMTLGDIVF KRISKRI
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications