missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FABP7 Protein

A cDNA sequence encoding the FABP7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11837-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978538

  • 137.00€ / 5µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FABP7 Protein
Research Use Only
Recombinant Protein
E. coli
MVEAFCATWK LTNSQNFDEY MKALGVGFAT RQVGNVTKPT VIISQEGDKV VIRTLSTFKN TEISFQLGEE FDETTADDRN CKSVVSLDGD KLVHIQKWDG KETNFVREIK DGKMVMTLTF GDVVAVRHYE KA
Greater than 95.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The FABP7 protein solution contains 25mM Tris-HCl pH 7.5, 2mM EDTA and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions